Learn More
1-Iodo-2-methylpropane, 97%, stab. with copper, Thermo Scientific Chemicals
33.90€ - 796.00€
Identificatori chimici
CAS | 513-38-2 |
---|---|
Formula molecolare | C4H9I |
Molecular Weight (g/mol) | 184.02 |
Numero MDL | MFCD00001084 |
InChI Key | BTUGGGLMQBJCBN-UHFFFAOYSA-N |
Sinonimo | isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane |
PubChem CID | 10556 |
IUPAC Name | 1-iodo-2-methylpropane |
SMILES | CC(C)CI |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11494160
|
Thermo Scientific Alfa Aesar
A15883.14 |
25 g |
33.90€
25g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11404170
|
Thermo Scientific Alfa Aesar
A15883.22 |
100 g |
110.00€
100g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11462598
|
Thermo Scientific Alfa Aesar
A15883.0B |
1 kg |
796.00€
1000g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.
Identificatori chimici
513-38-2 | |
184.02 | |
BTUGGGLMQBJCBN-UHFFFAOYSA-N | |
10556 | |
CC(C)CI |
C4H9I | |
MFCD00001084 | |
isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane | |
1-iodo-2-methylpropane |
Specifica
1-Iodo-2-methylpropane | |
513-38-2 | |
1.599 | |
12°C (53°F) | |
1.496 | |
UN2391 | |
Light sensitive | |
isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane | |
BTUGGGLMQBJCBN-UHFFFAOYSA-N | |
1-iodo-2-methylpropane | |
10556 | |
98% |
stabilized with copper | |
-93°C | |
120°C to 121°C | |
C4H9I | |
MFCD00001084 | |
1730927 | |
14,5144 | |
Insoluble in water. | |
CC(C)CI | |
184.02 | |
184.02 |
Sicurezza e movimentazione
GHS H Statement
H225-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H332-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: IODOMETHYLPROPANES
missing translation for 'einecsNumber' : 208-160-5
missing translation for 'rtecsNumber' : TZ4250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only