Learn More
13-cis-Retinoic acid
Descrizione
13-cis-Retinoic acid is used to inhibits cell proliferation and induces cell differentiation. 13-cis-Retinoic acid is primarily used in the treatment of severe recalcitrant nodular acne.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in chloroform, dimethyl sulfoxide, water, ethanol, and ether.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifica
Specifica
| Materiale o nome chimico | 13-cis-Retinoic acid |
| CAS | 4759-48-2 |
| Punto di fusione | 172°C to 175°C |
| Colore | Yellow to Orange |
| Formula molecolare | C20H28O2 |
| Numero MDL | MFCD00079542 |
| Quantità | 500 mg |
| Indice di Merck | 14,5228 |
| Sinonimo | Isotretinoin |
| Informazioni di solubilità | Soluble in chloroform,dimethyl sulfoxide,water,ethanol,and ether. |
| Vedi altri risultati |
RUO – Research Use Only
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.