Learn More
3-Aminothiophenol, 97%, Thermo Scientific Chemicals
Catalog Number p-7028256
Click to view available options
Quantità:
5 g
25 g
100 g
22948-02-3
C6H6NS
124.18
MFCD00007791
KFFUEVDMVNIOHA-UHFFFAOYSA-M
3-aminothiophenol, benzenethiol, 3-amino, 3-mercaptoaniline, m-amino thiophenol, 3-aminobenzene-1-thiol, 3-amino thiophenol, 3-amino-benzenethiol, pubchem2470, acmc-1co9c
31577
NC1=CC=CC([S-])=C1
22948-02-3
C6H6NS
124.18
MFCD00007791
KFFUEVDMVNIOHA-UHFFFAOYSA-M
3-aminothiophenol, benzenethiol, 3-amino, 3-mercaptoaniline, m-amino thiophenol, 3-aminobenzene-1-thiol, 3-amino thiophenol, 3-amino-benzenethiol, pubchem2470, acmc-1co9c
31577
NC1=CC=CC([S-])=C1
3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-Aminothiophenol
Densità 1.179
Punti di ebollizione 123°C to 125°C (8 mmHg)
Punto d'infiammabilità >100°C (212°F)
Indice di rifrazione 1.659
Quantità 5 g
Numero UN UN3267
Sensibilità Air sensitive
Informazioni di solubilità Soluble in water.
IUPAC Name 3-aminobenzenethiol
Peso formulazione 125.19
Percent Purity 97%