missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-L-phenylalanine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantità:
250 mg
1 g
Descrizione
It is applied in the facile removal of leader peptides from lanthipeptides by incorporation of a hydroxy acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied in the facile removal of leader peptides from lanthipeptides by incorporation of a hydroxy acid.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is applied in the facile removal of leader peptides from lanthipeptides by incorporation of a hydroxy acid.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Specifica
Materiale o nome chimico | 3-Bromo-L-phenylalanine |
Informazioni di solubilità | Slightly soluble in water. |
Peso formulazione | 244.09 |
Percent Purity | 95% |
Quantità | 250 mg |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'