missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxybenzonitrile, 95%, Thermo Scientific Chemicals

Codice prodotto. 11315469
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 2315-81-3
Formula molecolare C7H4ClNO
Molecular Weight (g/mol) 153.565
Numero MDL MFCD01567246
InChI Key CRYPJUOSZDQWJZ-UHFFFAOYSA-N
Sinonimo benzonitrile, 3-chloro-4-hydroxy, 2-chloro-4-cyanophenol, 3-chloro-4-hydroxy-benzonitrile, 3-chloro-4-hydroxybenzenecarbonitrile, pubchem4726, 4-cyano-2-chlorophenol, buttpark 4357-94, acmc-1clj1, pharmabridge p-1867, buttpark 43\57-94
PubChem CID 2735739
IUPAC Name 3-chloro-4-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1C#N)Cl)O

Specifica

Materiale o nome chimico 3-Chloro-4-hydroxybenzonitrile
Punto di fusione 147°C to 149°C
Quantità 1 g
Peso formulazione 153.57
Percent Purity 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'