Learn More
3-Methylpentane, 99+%, Thermo Scientific Chemicals
46.30€ - 162.00€
Identificatori chimici
CAS | 96-14-0 |
---|---|
Formula molecolare | C6H14 |
Molecular Weight (g/mol) | 86.18 |
Numero MDL | MFCD00009342 |
InChI Key | PFEOZHBOMNWTJB-UHFFFAOYSA-N |
Sinonimo | pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l |
PubChem CID | 7282 |
IUPAC Name | 3-methylpentane |
SMILES | CCC(C)CC |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11416720
|
Thermo Scientific Alfa Aesar
L03133.AE |
100 mL |
46.30€
100mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11426720
|
Thermo Scientific Alfa Aesar
L03133.AP |
500 mL |
162.00€
500mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
3-Methylpentane is used as a solvent in the preparation of vegetable oils, glues, coatings, and paints. It is also used in gasoline, rubber solvent, and petroleum ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methylpentane is used as a solvent in the preparation of vegetable oils, glues, coatings, and paints. It is also used in gasoline, rubber solvent, and petroleum ether.
Solubility
Soluble in water. 0.013 g/L at 20°C). Miscible in ether, acetone. Soluble in ethanol.
Notes
Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
Identificatori chimici
96-14-0 | |
86.18 | |
PFEOZHBOMNWTJB-UHFFFAOYSA-N | |
7282 | |
CCC(C)CC |
C6H14 | |
MFCD00009342 | |
pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l | |
3-methylpentane |
Specifica
3-Methylpentane | |
-118°C | |
63°C to 65°C | |
Petroleum-like | |
1.377 | |
UN1208 | |
pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l | |
PFEOZHBOMNWTJB-UHFFFAOYSA-N | |
3-methylpentane | |
7282 | |
≥99% |
96-14-0 | |
0.663 | |
−32°C (−26°F) | |
C6H14 | |
MFCD00009342 | |
1730734 | |
Soluble in water. 0.013g/L at 20°C). Miscible in ether,acetone. Soluble in ethanol. | |
CCC(C)CC | |
86.18 | |
86.18 |
Sicurezza e movimentazione
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
missing translation for 'einecsNumber' : 202-481-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only