Learn More
Hamilton™ 31 gauge, Kel-F Hub Needle, custom length (0.375 to 12 in), point style 2, 3, or 4, 6/PK
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. / 51 mm. Kel-F hub (KF) needles are available in convenient six packs. Custom lengths and point styles are available. These needles are available from 10 - 31 gauge.
Marca:  Hamilton™ 7750-22
Codice prodotto. 11510162
  • 259.00€ / pezzi 6
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Specification Sheet
Gauge31 gauge
Point Style2, 3, or 4
Needle LengthCustom Needle Length
AutoclavableYes
Gas SterilizableYes
Sales Quantity6 needles per pack
HubKel-F
Specifica
Specifica
31 G
Kel-F Hub Needle