Learn More
3M™ Protective Coverall 4565
L'indumento di protezione 3M™ 4565 ha caratteristiche di comfort desiderabili e fornisce una barriera resistente a determinate sostanze chimiche, spruzzi di liquidi e particolato.
Marca: 3M™ 7000089686
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Type 4/5/6 Coverall designed to offer a very good barrier to hazardous particles and limited liquid splash and spray
- Made of bicomponent nonwoven with ultra-low linting PE laminate for a noticeably comfortable drape
- 3 panel hood, 2-way zip and storm flap with sealable tape with elasticated back and ankles to reduce the risk of skin exposure
- Taped seams across the suit for increased hold out and strength reducing the risk of tearing and exposure
- Protection against EN 14126 Biological agents
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
- EN 1073-2 nuclear particulate hold out Til class 2