missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyphenoxy)aniline, 97%, Thermo Scientific™

Codice prodotto. 15485307
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 13066-01-8
Formula molecolare C13H13NO2
Molecular Weight (g/mol) 215.252
Numero MDL MFCD02663341
InChI Key XFOFRBMGVDBINH-UHFFFAOYSA-N
Sinonimo 4-2-methoxyphenoxy aniline, benzenamine, 4-2-methoxyphenoxy, 4-2-methoxyphenoxy phenylamine, 4-2-methoxyphenoxy-aniline, 4-2-methoxy-phenoxy-phenylamine
PubChem CID 1121067
IUPAC Name 4-(2-methoxyphenoxy)aniline
SMILES COC1=CC=CC=C1OC2=CC=C(C=C2)N

Specifica

Materiale o nome chimico 4-(2-Methoxyphenoxy)aniline
Quantità 1 g
Peso formulazione 215.25
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'