missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetanilide, 98%, Thermo Scientific Chemicals

Codice prodotto. 10327930
100 g, Bottiglia in vetro
Click to view available options
Quantità:
25 g
100 g
500 g
Imballaggio:
Bottiglia in vetro
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-88-8
Formula molecolare C8H8BrNO
Molecular Weight (g/mol) 214.06
Numero MDL MFCD00000092
InChI Key MSLICLMCQYQNPK-UHFFFAOYSA-N
Sinonimo 4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide
PubChem CID 7683
IUPAC Name N-(4-bromophenyl)acetamide
SMILES CC(=O)NC1=CC=C(Br)C=C1

Specifications

Materiale o nome chimico 4′-Bromoacetanilide
CAS min. % 97.5
CAS max % 100.0
Punto di fusione 166.0°C to 170.0°C
Colore Beige to White or Gray
Spettro a infrarossi Authentic
Intervallo percentuale saggio 97.5% min. (GC)
Imballaggio Bottiglia in vetro
Formula lineare CH3CONHC6H4Br
Quantità 100 g
Beilstein 12,642
Indice di Merck 15,1406
Informazioni di solubilità Solubility in water: practically insoluble. Other solubilities: sparingly soluble in hot water,soluble in benzene,chloroform and ethyl acetate,moderately soluble in alcohol
Peso formulazione 214.06
Percent Purity 98%
Forma fisica Crystalline Powder or Crystals
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Bromoacetanilide
Signal Word
  • Attenzione
Hazard Category
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Corrosione/irritazione della pelle Categoria 2
  • Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3
Hazard Statement
  • H319-Provoca grave irritazione oculare.
  • H335-Può irritare le vie respiratorie.
  • H315-Provoca irritazione cutanea.
Precautionary Statement
  • P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4'-Bromoacetanilide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.