Learn More
4-Fluoro-2-nitroaniline, 98%, Thermo Scientific Chemicals
115.00€ - 637.00€
Identificatori chimici
CAS | 364-78-3 |
---|---|
Formula molecolare | C6H5FN2O2 |
Molecular Weight (g/mol) | 156.12 |
Numero MDL | MFCD00007830 |
InChI Key | PUGDHSSOXPHLPT-UHFFFAOYSA-N |
Sinonimo | 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421 |
PubChem CID | 67769 |
IUPAC Name | 4-fluoro-2-nitroaniline |
SMILES | NC1=CC=C(F)C=C1[N+]([O-])=O |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11364729
|
Thermo Scientific Alfa Aesar
A10685.18 |
50 g |
115.00€
50g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11433454
|
Thermo Scientific Alfa Aesar
A10685.22 |
100 g |
180.00€
100g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11443454
|
Thermo Scientific Alfa Aesar
A10685.36 |
500 g |
637.00€
500g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases and strong acids.
Identificatori chimici
364-78-3 | |
156.12 | |
PUGDHSSOXPHLPT-UHFFFAOYSA-N | |
67769 | |
NC1=CC=C(F)C=C1[N+]([O-])=O |
C6H5FN2O2 | |
MFCD00007830 | |
2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421 | |
4-fluoro-2-nitroaniline |
Specifica
4-Fluoro-2-nitroaniline | |
90°C to 94°C | |
C6H5FN2O2 | |
2210197 | |
Insoluble in water. | |
NC1=CC=C(F)C=C1[N+]([O-])=O | |
156.12 | |
156.12 |
364-78-3 | |
89°C (192°F) | |
MFCD00007830 | |
2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421 | |
PUGDHSSOXPHLPT-UHFFFAOYSA-N | |
4-fluoro-2-nitroaniline | |
67769 | |
98% |
Sicurezza e movimentazione
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H312+H332-H315-H319-H335
missing translation for 'einecsNumber' : 206-666-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only