Learn More
4-Methoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals
31.30€ - 302.00€
Identificatori chimici
CAS | 2284-20-0 |
---|---|
Formula molecolare | C8H7NOS |
Molecular Weight (g/mol) | 165.21 |
Numero MDL | MFCD00011676 |
InChI Key | VRPQCVLBOZOYCG-UHFFFAOYSA-N |
Sinonimo | 4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate |
PubChem CID | 75293 |
IUPAC Name | 1-isothiocyanato-4-methoxybenzene |
SMILES | COC1=CC=C(C=C1)N=C=S |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11394699
|
Thermo Scientific Alfa Aesar
A15331.04 |
2 g |
31.30€
2g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11448236
|
Thermo Scientific Alfa Aesar
A15331.09 |
10 g |
90.60€
10g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11458236
|
Thermo Scientific Alfa Aesar
A15331.18 |
50 g |
302.00€
50g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
Identificatori chimici
2284-20-0 | |
165.21 | |
VRPQCVLBOZOYCG-UHFFFAOYSA-N | |
75293 | |
COC1=CC=C(C=C1)N=C=S |
C8H7NOS | |
MFCD00011676 | |
4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate | |
1-isothiocyanato-4-methoxybenzene |
Specifica
4-Methoxyphenyl isothiocyanate | |
18°C to 20°C | |
279°C to 281°C | |
Pungent | |
1.6489 | |
UN2922 | |
Moisture sensitive | |
It hydrolyzes in water. | |
COC1=CC=C(C=C1)N=C=S | |
165.21 | |
165.22 |
2284-20-0 | |
1.196 | |
>110°C (230°F) | |
C8H7NOS | |
MFCD00011676 | |
606967 | |
4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate | |
VRPQCVLBOZOYCG-UHFFFAOYSA-N | |
1-isothiocyanato-4-methoxybenzene | |
75293 | |
98% |
Sicurezza e movimentazione
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 218-921-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only