missing translation for 'onlineSavingsMsg'
Learn More

4-n-Decylaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7044889
Click to view available options
Quantità:
1 g
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
37529-30-9
C16H27N
233.399
MFCD00007918
WGENWPANMZLPIH-UHFFFAOYSA-N
4-n-decylaniline, p-decylaniline, benzenamine, 4-decyl, p-n-decylaniline, 4-decylphenylamine, 4-decyphenylamine, 4-decyl-phenylamine, 4-decylaniline, ksc225q8h
92309
4-decylaniline
CCCCCCCCCCC1=CC=C(C=C1)N
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
37529-30-9
C16H27N
233.399
MFCD00007918
WGENWPANMZLPIH-UHFFFAOYSA-N
4-n-decylaniline, p-decylaniline, benzenamine, 4-decyl, p-n-decylaniline, 4-decylphenylamine, 4-decyphenylamine, 4-decyl-phenylamine, 4-decylaniline, ksc225q8h
92309
4-decylaniline
CCCCCCCCCCC1=CC=C(C=C1)N

It is used to synthesize p,p'-Di-n-decyl-azobenzol could be obtained by the reactant of 4-Decylaniline, and the reagents of O2, CuCl, pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to synthesize p,p′-Di-n-decyl-azobenzol could be obtained by the reactant of 4-Decylaniline, and the reagents of O2, CuCl, pyridine.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away air and from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-n-Decylaniline
Punto di fusione 23°C to 25°C
Punti di ebollizione 218°C to 220°C (14 mmHg)
Punto d'infiammabilità >110°C (230°F)
Indice di rifrazione 1.51
Quantità 1 g
Beilstein 2721403
Sensibilità Air sensitive
Informazioni di solubilità Insoluble in water.
Peso formulazione 233.4
Percent Purity 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'