Learn More
5-Methyl-1-hexyne, 98%, Thermo Scientific Chemicals
49.40€ - 186.00€
Identificatori chimici
CAS | 2203-80-7 |
---|---|
Formula molecolare | C7H12 |
Molecular Weight (g/mol) | 96.173 |
Numero MDL | MFCD00041610 |
InChI Key | HKNANEMUCJGPMS-UHFFFAOYSA-N |
Sinonimo | 5-methyl-1-hexyne, 1-hexyne, 5-methyl, isoheptyne, unii-lro6ea6hk1, lro6ea6hk1, isoamylacetylene, isohept-1-yne, acmc-1cg6t, hknanemucjgpms-uhfffaoysa |
PubChem CID | 137490 |
IUPAC Name | 5-methylhex-1-yne |
SMILES | CC(C)CCC#C |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11372326
|
Thermo Scientific Alfa Aesar
L06014.06 |
5 g |
49.40€
5g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11382326
|
Thermo Scientific Alfa Aesar
L06014.14 |
25 g |
186.00€
25g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
5-Methyl-1-hexyne is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Methyl-1-hexyne is used as pharmaceutical intermediates.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Identificatori chimici
2203-80-7 | |
96.173 | |
HKNANEMUCJGPMS-UHFFFAOYSA-N | |
137490 | |
CC(C)CCC#C |
C7H12 | |
MFCD00041610 | |
5-methyl-1-hexyne, 1-hexyne, 5-methyl, isoheptyne, unii-lro6ea6hk1, lro6ea6hk1, isoamylacetylene, isohept-1-yne, acmc-1cg6t, hknanemucjgpms-uhfffaoysa | |
5-methylhex-1-yne |
Specifica
5-Methyl-1-hexyne | |
0.732 | |
−4°C (24°F) | |
1.4075 | |
UN3295 | |
5-methyl-1-hexyne, 1-hexyne, 5-methyl, isoheptyne, unii-lro6ea6hk1, lro6ea6hk1, isoamylacetylene, isohept-1-yne, acmc-1cg6t, hknanemucjgpms-uhfffaoysa | |
HKNANEMUCJGPMS-UHFFFAOYSA-N | |
5-methylhex-1-yne | |
137490 | |
98% |
2203-80-7 | |
91°C to 93°C | |
C7H12 | |
MFCD00041610 | |
1731845 | |
Not miscible in water. | |
CC(C)CCC#C | |
96.173 | |
96.17 |
Sicurezza e movimentazione
GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H225-H304
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only