Learn More
uvex 9193 Sportstyle Safety Spectacles
Codice prodotto. p-7221900 Sfoglia Tutto Uvex Prodotti
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Lens Tint:
Clear
Gray
Light Brown
Light Green
Silver Mirror
Colore telaio:
Black, Anthracite
Black, Blue
Black, Gray
Black, Lime Green
Black, White
White, Black
Dimensione della confezione:
Pezzo
Codice prodotto. 17237291
Marca: uvex 9193885
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Innovative safety spectacles featuring a sporty design, combined with maximum functionality and safety. Feather-light construction weighs only 23 grams, making these spectacles comfortable to wear for extended periods of time.
  • Innovative safety spectacles feature a sporty design that combines functionality and safety 
  • Feather-light construction weighs only 23 grams
  • Soft, adjustable nosepiece with soft, anti-slip earpieces for a pressure-free fit
  • Lens geometry ensures optimum vision
Supravision AR Coating
  • Inside: Anti-reflective
  • Outside: Anti-reflective
  • Ideal for working conditions with strong or mixed lighting conditions
Supravision Excellence Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
Supravision Extreme Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant
  • Ideal for working areas with long-term exposure to condensation, and high levels of dirt
Supravision Plus Coating
  • Inside: Anti-fog and scratch-resistant
  • Outside: Anti-fog and scratch-resistant
  • Suitable for environments with frequent temperature and humidity fluctuations
Supravision Variomatic Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant
  • Self-coloring filters automatically darken within 10 seconds and brighten after 30 seconds
  • Suitable for working areas with frequent movement from inside to outside, and from light to dark
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità EN 166:2001, EN 172:1994 + A1:2000 + A2:2001
Lens Coating Anti-fog
Lens Tint Silver Mirror
Product Type Ergonomic Safety Glasses
Colore telaio Black, Gray
Modello 9193 Sportstyle
Valore di trasmittanza 0.12
Peso (sistema metrico) 23 g
Materiale Polycarbonate
Monouso Reusable
Lens Material Policarbonato
Materiale del telaio Plastic
UV Protection UV400
Frame Type Standard
Labeling W 166 FT CE - 5-3,1 W1 FTN CE