Learn More
Fisherbrand™ Piastre per PCR da 96 pozzetti a basso profilo con bordo
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Colore:
Blue
Green
Incolore
Red
Yellow
Dimensione della confezione:
pezzi 25
Descrizione
Fit most thermal cyclers
- Virgin polypropylene
- Clear well bottom for sample visibility
- Stackable
- Raised-rim design
- Sealable using adhesive films and foils, heat seals, and cap strips
- 200μL maximum well volume when used with adhesive and heat seals
- Cut-away corner and alphanumeric grid
- SBS (Society for Biomolecular Screening) footprint
- Automation friendly
- Eight holes in the skirt aid plate positioning and removal from thermal cycler block
- Cleanroom produced
Specifica
Specifica
| Formato | Standard |
| Matrice | 8 x 12 |
| Colore | Red |
| Product Type | PCR Microplate |
| Skirt Style | Skirted |
| Materiale | Polypropylene |
| Volume (sistema metrico) pozzetto | 200 μL |
| No. of Wells | 96 |
| Da utilizzare con (applicazione) | PCR, RT-PCR applications |
| Con bordo | Yes |
| Vedi altri risultati |