missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IgM (Dog) ELISA Kit
Enzyme-linked immunoassay for the quantitative determination of IgM in Dog biological samples
Specifica
Metodo di rilevamento | Colorimetric |
---|---|
Coniugato | HRP |
Cross-reattività | Dog |
Product Type | IgM (Dog) ELISA kit |
Quantità | 1 Kit |
Specifica
Colorimetric | |
Dog | |
1 Kit | |
100 μL |
HRP | |
IgM (Dog) ELISA kit | |
For research use only (RUO) | |
Store the kit at 4°C |