missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Product Type:
1536-well Adapter for 10 mm Plate w/o Lid
384-well Adapter for Plate w/ Lid
384-well Adapter for Plate w/o Lid
6-48-well Adapter for Plate w/ Lid
6-48-well Adapter for Plate w/o Lid
96-well Adapter for Plate w/ Lid
96-well Adapter for Plate w/o Lid
96-well adapter for PCR plate without lid
Aspirate tube assembly for a dispenser
Dispenser syringe, 1 mL
Dispenser syringe, 5 mL
Dispensing tube assembly for a dispenser
Emission Filter
Emission Filter Pick-up Tool
Liquid Protection Tray
Microplate package for Varioskan LUX IQ/OQ
Robotic plate tray adapter for plates without lid
Robotic plate tray for plates with and without lids
Storage Holder for Plate Adapters
Universal plate tray for plates without lid
Upgrade Kit for Luminometry and TRF
Upgrade kit for luminometry and AlphaScreen
Upgrade kit for luminometry, AlphaScreen and TRF
Upgrade kit for two dispensers
Dimensione della confezione:
Pezzo
Descrizione
Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.
Varioskan LUX is a modular instrument, you can upgrade it with missing features later.
If the dispensers are in frequent use, it is recommended to replace the aspirate and dispensing tubings once a year.
- Installation of the upgrade kits must be carried out by trained and authorized service personnel.
- For TRF and/or AlphaScreen technology upgrades, the instrument must be equipped with a luminescence module.
- TRF and/or AlphaScreen technology upgrades include excitation filters but not emission filters. Please order emission filters separately.
- Please contact your local representative to discuss about the instrument upgrade option
- Adapters adjust the plates to the optimum height for measuring and dispensing.
- Choose the adapter based on the plate format in use and whether the lid is needed or not.
- The universal plate tray is for basic use and it is compatible with all plate formats (6- to 1536-well plates).
- A robotic plate tray is for automated use with robots and is compatible with 96- to 1536-well plate formats.
If the dispensers are in frequent use, it is recommended to replace the aspirate and dispensing tubings once a year.
Specifica
Specifica
| Product Type | Microplate package for Varioskan LUX IQ/OQ |
| Da utilizzare con (apparecchiatura) | Varioskan LUX Microplate Reader |
| Tipo di piastra | 384-well Plate |
| Linea di prodotti | Varioskan |
| Da utilizzare con (applicazione) | Varioskan LUX performance qualification |
| Include | Microplates needed for Varioskan LUX qualification |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto