missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader

Codice prodotto. 17954312 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Product Type:
1536-well Adapter for 10 mm Plate w/o Lid
384-well Adapter for Plate w/ Lid
384-well Adapter for Plate w/o Lid
6-48-well Adapter for Plate w/ Lid
6-48-well Adapter for Plate w/o Lid
96-well Adapter for Plate w/ Lid
96-well Adapter for Plate w/o Lid
96-well adapter for PCR plate without lid
Aspirate tube assembly for a dispenser
Dispenser syringe, 1 mL
Dispenser syringe, 5 mL
Dispensing tube assembly for a dispenser
Emission Filter
Emission Filter Pick-up Tool
Liquid Protection Tray
Microplate package for Varioskan LUX IQ/OQ
Robotic plate tray adapter for plates without lid
Robotic plate tray for plates with and without lids
Storage Holder for Plate Adapters
Universal plate tray for plates without lid
Upgrade Kit for Luminometry and TRF
Upgrade kit for luminometry and AlphaScreen
Upgrade kit for luminometry, AlphaScreen and TRF
Upgrade kit for two dispensers
Dimensione della confezione:
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 17954312

Marca: Thermo Scientific™ N16308S

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader.

Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.

Varioskan LUX is a modular instrument, you can upgrade it with missing features later.
  • Installation of the upgrade kits must be carried out by trained and authorized service personnel.
  • For TRF and/or AlphaScreen technology upgrades, the instrument must be equipped with a luminescence module.
  • TRF and/or AlphaScreen technology upgrades include excitation filters but not emission filters. Please order emission filters separately.
  • Please contact your local representative to discuss about the instrument upgrade option
Plate adapters are used for adapting plates that are of different heights.
  • Adapters adjust the plates to the optimum height for measuring and dispensing.
  • Choose the adapter based on the plate format in use and whether the lid is needed or not.
There are two types of plate trays: universal and robotic.
  • The universal plate tray is for basic use and it is compatible with all plate formats (6- to 1536-well plates).
  • A robotic plate tray is for automated use with robots and is compatible with 96- to 1536-well plate formats.

If the dispensers are in frequent use, it is recommended to replace the aspirate and dispensing tubings once a year.
TRUSTED_SUSTAINABILITY

Specifica

Product Type Liquid Protection Tray
Da utilizzare con (apparecchiatura) Varioskan LUX Multimode Microplate Reader
Linea di prodotti Varioskan
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato