missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals

Codice prodotto. 11384358
Click to view available options
Quantità:
250 mL
1 L
4 L
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Water <100ppm, typically 50ppm (0.005%)

Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 75-05-8
Formula molecolare C2H3N
Molecular Weight (g/mol) 41.053
Numero MDL MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Sinonimo methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifica

Materiale o nome chimico Acetonitrile
Punto di fusione -46°C / -50.8°F
Colore Colorless
Aspetto Colorless liquid
Densità 0.786
Punti di ebollizione 80°C to 82°C
Quantità 4 L
Punto d'infiammabilità 5°C (41°F)
Punto di combustione 525°C (977°F)
Forma fisica Liquid
Indice di rifrazione 1.344
Intervallo percentuale saggio >99.8%
Pressione del vapore 97 hPa (73mm Hg) at 20°C
Numero UN 1648;Colorless
Beilstein 741857
Informazioni di solubilità Miscible with water,alcohols,ethers,acetone,chloroform,carbon tetrachloride and ethylene chloride. Immiscible with saturated hydrocarbons (petroleum fractions).
Peso formulazione 41.05
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'