missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylenedicarboxylic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantità:
5 g
25 g
100 g
Descrizione
Acetylenedicarboxylic acid is employed in the preparation of dimethyl acetylenedicarboxylate and potassium hydrogenacetylenedicarboxylate. It is also used as a laboratory reagent. Further, it is used as a precursor in organic synthesis and also serves as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acetylenedicarboxylic acid is employed in the preparation of dimethyl acetylenedicarboxylate and potassium hydrogenacetylenedicarboxylate. It is also used as a laboratory reagent. Further, it is used as a precursor in organic synthesis and also serves as a pharmaceutical intermediate.
Solubility
Soluble in diethyl ether and water.
Notes
Air sensitive. Store in a cool place. Incompatible with bases, alkali, reducing agents and oxidizing agents.
Acetylenedicarboxylic acid is employed in the preparation of dimethyl acetylenedicarboxylate and potassium hydrogenacetylenedicarboxylate. It is also used as a laboratory reagent. Further, it is used as a precursor in organic synthesis and also serves as a pharmaceutical intermediate.
Solubility
Soluble in diethyl ether and water.
Notes
Air sensitive. Store in a cool place. Incompatible with bases, alkali, reducing agents and oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Materiale o nome chimico | Acetylenedicarboxylic acid |
Punto di fusione | ∼182°C (decomposition) |
Punto d'infiammabilità | 131°C (267°F) |
Quantità | 5 g |
Numero UN | UN2928 |
Beilstein | 878357 |
Informazioni di solubilità | Soluble in diethyl ether and water. |
Peso formulazione | 114.06 |
Percent Purity | 97% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'