Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Glass Fiber Filter
Available with GxF multi-layered glass fiber prefilter, providing two to four times the throughput of standard glass fiber prefilter devices, and allowing quick and easy filtration of your most difficult-to-filter samples.
Marca: Cytiva (formerly Pall Lab) 4523
101.72 EUR valido fino al 2024-12-31
Usa il codice promo "21934" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.43800kg
Descrizione
- For research use only
Specifica
Borosilicate Glass Fiber | |
Non-sterile | |
200/Cs. | |
25 mm |
1 μm | |
Polypropylene | |
50/Pk. |