Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Glass Fiber Filter
Available with GxF multi-layered glass fiber prefilter, providing two to four times the throughput of standard glass fiber prefilter devices, and allowing quick and easy filtration of your most difficult-to-filter samples.
Marca: Cytiva (formerly Pall Lab) 4524
Dettagli aggiuntivi : Peso : 0.98800kg
Descrizione
- For research use only
Specifica
Borosilicate Glass Fiber | |
Non-sterile | |
60/Cs. | |
37 mm |
1 μm | |
Polypropylene | |
15/Pk. |