Learn More
Grant Instruments™ TX150 Series Heated Circulating Bath, Stainless Steel Tank
Click to view available options
Depth (Metric) Interior:
300 mm
325 mm
505 mm
690 mm
Altezza (sistema metrico) interno:
150 mm
200 mm
Larghezza (sistema metrico) interno:
150 mm
300 mm
Gamma di temperatura:
-15°C to +200°C
0°C to 200°C
Dimensione della confezione:
Pezzo
Descrizione
- Stability ±0.01ºC
- Large, bright full colour display
- Icon driven home screen via a dial and two function buttons
- Programming/temperature profiling (TX150, 1 program with 30 segments)
- Model available with/without clamp (T-Clamp)
- Low-liquid detection (float switch)
- 5 point user calibration
- User adjustable over temperature dial
- 5 point user calibration
Specifica
Specifica
| Gamma di temperatura | -15°C to +200°C |
| Include | Includes TX150 immersion heater/stirrer with ST26 Bath |
| Materiale | Stainless Steel |
| Larghezza (sistema metrico) interno | 300 mm |
| Altezza (sistema metrico) interno | 200 mm |
| Depth (Metric) Interior | 505 mm |