missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose MS8, molecular sieve grade, for small DNA fragments

For small DNA fragments

153.77€ - 375.87€

Identificadores de productos químicos

CAS 9012-36-6
Numero MDL MFCD00081294
Ver más especificaciones

Productos 2
Código de producto Marca Quantità Precio Cantidad y disponibilidad  
Código de producto Marca Quantità Precio Cantidad y disponibilidad  
15447615
missing translation for 'viewDocuments' Detalles de la promoción
Thermo Scientific Chemicals
H26738.14
25 g
153.77€
25g
In stock
per visualizzare lo stock disponibile
Añadir a la cesta
 
11332729
missing translation for 'viewDocuments' Detalles de la promoción
Thermo Scientific Chemicals
H26738.22
100 g
375.87€
100g
Spedizione stimata: 20-11-2024
per visualizzare lo stock disponibile
Añadir a la cesta
 
Descripción

Descripción

Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Especificaciones

Identificadores de productos químicos

9012-36-6
MFCD00081294

Especificaciones

Agarose MS8
9012-36-6
Soluble in water.
Hygroscopic; Moisture sensitive
for small DNA fragments
MFCD00081294
Molecular Sieve
Videos
Seguridad y manipulación

Seguridad y manipulación

missing translation for 'einecsNumber' : 232-731-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documentos

Documentos

Certificados

RUO – Research Use Only