Learn More
Abeba™ Air-Cushion 32300 Shoes
Air cushion PU-sole with high slip resistance. Sanitized™ treated.
Marca: Abeba™ 32300/41
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- CE EN ISO 20347:2012 O1 SRA
- Smooth leather, perforated
- White
- Slip resistance PU outsole sole
- Replaceable insole (article no. 3551)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with elastic band
Specifica
CE EN ISO 20347:2012 O1 SRA | |
Microfiber | |
41 | |
Polyurethane |
White | |
Leather | |
Slip-resistant | |
32300 |