Learn More
Aalborg™ Air Direct Glass Flow Tube 150mm Reference Scale
Codice prodotto. 11718368
Click to view available options
Velocità di flusso dell'aria:
1.25 l/min.;;en;
10 L/min.
100 mL/min.
1093 mL/min.
143 mL/min.
1637 mL/min.
19 mL/min.
2.5 L/min.
200 mL/min.
23 L/min.
25 mL/min.
264 mL/min.
30 L/min.
300 ml/min.;;en;
374 mL/min.
42 L/min.
49 mL/min.
50 mL/min.
500 mL/min.
5188 mL/min.
70 mL/min.
75 mL/min.
8505 mL/min.
Dimensione della confezione:
Pezzo
Codice prodotto. 11718368
Marca: Aalborg™ FT03439STVA
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Aalborg™ Air Direct Glass Flow Tube Reference Scale reults in superior mL and L readings by providing a scale of meaurements in a vairety of volumes.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifica
Tipo Air Direct Flow Tube
Larghezza (sistema metrico) 150 mm
Velocità di flusso dell'aria 1637 mL/min.