missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ AKR7A2 Recombinant Protein

Marca:  Abnova™ H00008574-P01.25ug

 vedi altre versioni di questo prodotto

Codice prodotto. 16172622

  • 521.00€ / 25µg

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH04111.3
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
61.82
8
Glutathione Sepharose 4 Fast Flow
25 μg
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AKR7A2
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
8574
AKR7A2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
AFAR/AFAR1/AFB1-AR1/AKR7
AKR7A2
Wheat Germ (in vitro)
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.