missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin Antibody (DM1A) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 89 publications
236.00€ - 625.00€
Specifica
| Antigene | alpha Tubulin |
|---|---|
| Clone | DM1A |
| Concentrazione | 1.0 mg/mL |
| Diluizione | Western Blot 1:5000, Simple Western 1:50, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50000-1:100000, Immunoprecipitation 1:50-1:100, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, Immunomicroscopy, Flow (Intracellular), CyTOF-ready |
| Applicazioni | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunomicroscopy, CyTOF |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18759223
|
Novus Biologicals
NB100-690SS |
0.025 mL |
236.00€
25µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18073995
|
Novus Biologicals
NB100-690 |
0.1 mL |
625.00€
0.10mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Fungi, Guinea Pig, Goat, Hamster, Parasite, Monkey, Primate, Rabbit, Xenopus, Yeast samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunomicroscopy, Flow (Intracellular), CyTOF-ready.Specifica
| alpha Tubulin | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunomicroscopy, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
| alpha tubulin loading control, alpha tubulin monoclonal, Alpha-tubulin 3, B-ALPHA-1, DM1A, DM1A anti-tubulin, dm1a monoclonal, DM1a tubulin, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA1 monoclonal, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin monoclonal, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| TUBA1A | |
| IgG1 κ | |
| Protein G purified | |
| This alpha Tubulin Antibody (DM1A) does not cross-react with beta Tubulin. |
| DM1A | |
| Western Blot 1:5000, Simple Western 1:50, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50000-1:100000, Immunoprecipitation 1:50-1:100, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, Immunomicroscopy, Flow (Intracellular), CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Fungi, Guinea Pig, Goat, Hamster, Parasite, Monkey, Primate, Rabbit, Xenopus species, Yeast | |
| 7846 | |
| This alpha Tubulin Antibody (DM1A) was developed against native chicken brain microtubules. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 50 kDa |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto