Learn More
Ansell AlphaTec™ 2000 Ts PLUS
434.11 EUR valido fino al 2024-12-31
Usa il codice promo "22917" per avere il tuo prezzo promozionale.
Usa il codice promo "22917" per avere il tuo prezzo promozionale.
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Descrizione
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone free, critical in spray painting applications
Specifica
Size | Small |
Materiale | Microporous Non-woven Polyethylene Laminate |
Colore | White |
Tipo di giuntura | Stitched, Taped |
Certifications/Compliance (Europe) | EN 374:2003, , EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003, DIN 32781:2010 |
Include | Coverall with Collar |
Da utilizzare con (applicazione) | Body Painting Including Surface Inspection, Cleaning of Plant and Machines, Fibre Glass/Resin Applications/Ceramic Fibres, Product Inspection, Loading and Hauling |
Peso (sistema metrico) | 63 gsm |
Stile | W20T103 |