Learn More
Ansell™ AlphaTec 53-001 Gloves
Chemical protection gloves with nitrile neoprene nitrile chemical gauntlet and Ansell Grip technology
Marca: Ansell™ 53001060
11.45 EUR valido fino al 2024-07-31
Usa il codice promo "21640" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 7.63200kg
Descrizione
- These acid-resistant gloves offer a nitrile and neoprene barrier that provides advanced chemical protection from acids to hydrocarbons and beyond
- These AlphaTec™ 53-001 black chemical resistant gloves have ‘Type A’ chemical protection under EN ISO 374:2016
- MICROCHEM™ Chemical Barrier Technology combines polymers in proprietary blends or multi-layer structures specific to their application, providing superior protection to wearers
- ANSELL GRIP™ Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
Specifica
White, Black | |
Nylon | |
53-001 |
Yes | |
6 | |
350 mm |