Learn More
Cytiva Nucleotidi fluorescenti Amersham™ CyDye™
Sfoglia Tutto Cytiva Prodotti
Click to view available options
Product Type:
Cy3-CTP
Cy3-UTP
Cy3-dCTP
Cy3-dCTP, Cy5-dCTP
Cy3-dUTP
Cy3-dUTP, Cy5-dUTP
Cy3.5-dCTP
Cy5-CTP
Cy5-UTP
Cy5-dCTP
Cy5-dUTP
Cy5.5-dCTP
Quantità:
100 nm
100 nmol
25 nmol
250 nmol
5 x 25 nmol
Colore:
Arancione
Far-Red
Near IR
Orange
Orange, Far-Red
Scarlet
Dimensione della confezione:
1 set
Pezzo
Descrizione
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.
- Low nonspecific binding
- High photostability
- High water solubility
- pH insensitive
Specifica
Specifica
| Descrizione | CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy dye labelled nucleotides exhibit the following properties: |
| Product Type | Cy5-dCTP |
| Reagenti | Fluorescent Labeling Reagents |
| Quantità | 250 nmol |
| Colore | Far-Red |