missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium dihydrogen phosphate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
33.00€ - 277.00€
Identificatori chimici
CAS | 7722-76-1 |
---|---|
Formula molecolare | H6NO4P |
Molecular Weight (g/mol) | 115.03 |
Numero MDL | MFCD00003396 |
InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
Sinonimo | ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate |
PubChem CID | 24402 |
ChEBI | CHEBI:62982 |
SMILES | N.OP(O)(O)=O |
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
10358570
|
Thermo Scientific Acros
193701000 |
100 g | Plastic bottle |
33.00€
100g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
10781851
|
Thermo Scientific Acros
193705000 |
500 g | Plastic bottle |
84.50€
500g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
11974291
|
Thermo Scientific Acros
193700025 |
2.5 kg | Plastic bottle |
277.00€
2.50kg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
24402 | |
N.OP(O)(O)=O |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
CHEBI:62982 |
Specifica
Ammonium dihydrogen phosphate | |
7722-76-1 | |
Crystalline Powder | |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
azanium;dihydrogen phosphate | |
24402 | |
115.03 | |
Trace Metal Basis |
190.0°C | |
White | |
99.90% | |
NH4H2PO4 | |
15,541 | |
Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone | |
N.OP(O)(O)=O | |
115.03 | |
CHEBI:62982 | |
99.9% |
Sicurezza e movimentazione
missing translation for 'einecsNumber' : 231-764-5
RUO – Research Use Only