Learn More
Phenolics Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Marca: N/A CLPS-A
Dettagli aggiuntivi : Peso : 0.00100kg Transport : UN number : 1593 Chem class : 6.1 Pack group : III
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Descrizione
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
2,4,6-Trichlorophenol | 88-06-2 | 2000μg/mL |
2,4-Dichlorophenol | 120-83-2 | 2000μg/mL |
2,4-Dimethylphenol | 105-67-9 | 2000μg/mL |
2,4-Dinitrophenol | 51-28-5 | 2000μg/mL |
2-Chlorophenol | 95-57-8 | 2000μg/mL |
2-Nitrophenol | 88-75-5 | 2000μg/mL |
4,6-Dinitro-2-methylphenol | 534-52-1 | 2000μg/mL |
4-Chloro-3-methylphenol | 59-50-7 | 2000μg/mL |
4-Nitrophenol | 100-02-7 | 2000μg/mL |
Pentachlorophenol | 87-86-5 | 2000μg/mL |
Phenol | 108-95-2 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifica
Phenolics Mix | |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
For US EPA Methods 604, 625, 8270 |
USEPA Methods 604, 625, SW-846 Method 8270 | |
1 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
MeCl |
Sicurezza e movimentazione
- Pericolo
Product intended for laboratory use only