missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abeba™ Anatom 32281 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
131.00€
Specifica
Certificazione/Conformità | CE EN ISO 20345:2011 S3 SRC |
---|---|
Colore | Black |
Materiale | Microfiber |
Caratteristiche suola | Slip-resistant |
Materiale suola | Polyurethane |
Codice del prodotto | Marca | Size | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Size | Prezzo | Quantità e disponibilità | |||||
15699607
|
Abeba™
32281/36 |
36 | N/A | ||||||
15609617
|
Abeba™
32281/37 |
37 | N/A | ||||||
15619617
|
Abeba™
32281/38 |
38 | N/A | ||||||
15629617
|
Abeba™
32281/39 |
39 | N/A | ||||||
15639617
|
Abeba™
32281/40 |
40 | N/A | ||||||
15649617
|
Abeba™
32281/41 |
41 | N/A | ||||||
15659617
|
Abeba™
32281/42 |
42 | N/A | ||||||
15669617
|
Abeba™
32281/43 |
43 | N/A | ||||||
15679617
|
Abeba™
32281/44 |
44 | N/A | ||||||
15689617
|
Abeba™
32281/45 |
45 | N/A | ||||||
15699617
|
Abeba™
32281/46 |
46 | N/A | ||||||
15609627
|
Abeba™
32281/47 |
47 | N/A | ||||||
15619627
|
Abeba™
32281/48 |
48 | N/A | ||||||
Descrizione
- CE EN ISO 20345:2011 S3 SRC
- Microfiber black
- Steel toe cap
- Shock absorbant PU midsole
- Metalfree penetration resistance
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Cold insulating lining
- Adjustable instep area
- velcro fastening
- Reflective material
- Suitable for heel and foot lift
- Cushioned ankle support
Specifica
CE EN ISO 20345:2011 S3 SRC | |
Microfiber | |
Polyurethane | |
Steel |
Black | |
Slip-resistant | |
32281 |