Learn More
ABEBA™ Anatom Black/Red Safety Shoes S3
Robust, durable, black/red water and tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
Marca: ABEBA™ 2238/41
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Comfort, robustness, durability
- Comply to EN ISO 20345: 2012, S3, SRC
- Slip resistant PU double density outsole
- Black/red water and tear resistant Cordura
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Scratch-resistant coated fabric in toe and heel area
- Breathable and antibacterial lining with silver ions
- Removable active comfort insole
Specifica
EN ISO 20345: 2012, S3, SRC | |
Cordura | |
Steel | |
Low-cut | |
S3 SRC |
Black, Red | |
41 | |
Men's | |
Yes |