missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-ADAM12 Matched Antibody Pair, Abnova™
Marca: Abnova H00008038-AP51.1Set
Includes: Capture antibody: mouse monoclonal anti-ADAM12 (100μg); Detection antibody: rabbit purified polyclonal anti-ADAM12 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Descrizione
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ADAM12
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair
Specifica
ADAM12 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ADAM12. | |
MCMP/MCMPMltna/MLTN/MLTNA | |
Mouse, Rabbit | |
RUO | |
8038 | |
Human | |
Purified |
ELISA | |
Unconjugated | |
ADAM12 | |
ADAM12 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-ADAM12, IgG1 Kappa (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-ADAM12 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto