missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-AK1 Ab-3 Matched Antibody Pair, Abnova™
Marca: Abnova H00000203-AP13.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-AK1 (100μg); Detection antibody: mouse monoclonal anti-AK1, IgG1 Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
Host Species: Rabbit, Mouse
Isotype: IgG, IgG1, κ
Cross Reactivity: Human
ELISA Pair
Specifica
AK1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human AK1. | |
AK1 | |
Affinity Purified | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-AK1 (100 μg) 2. Detection antibody: mouse monoclonal anti-AK1, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
AK1 | |
Mouse, Rabbit | |
1 Set | |
203 | |
Human | |
IgG, IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto