missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM8 Mouse anti-Human, Clone: GM2H6, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA117768
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
This antibody recognizes native human CEACAM8 protein expressed in transfected mammalian cells.
CEA-related cell adhesion molecule 8 (CEACAM8, CD66b) belongs to the carcinoembryonic antigen (CEA) gene family. It encodes a glycosylphosphatidylinositol (GPI)-linked glycoprotein with a MW of 95kDa which is expressed in cells of the granulocyte-lineage. It is expressed in neutrophils and eosinophils and is characterized as a granulocyte-specific activation antigen. CEACAM8 could serve as a marker for granulocyte activities. Like all members of the CEA family, it consists of a single N domain, with structural homology to the immunoglobulin variable domains, followed by two immunoglobulin constant-like A and B domains.Specifica
CEACAM8 | |
Monoclonal | |
2 mg/mL | |
CEACAM8 | |
CD66b, CD67, CGM6, NCA-95 | |
Mouse | |
Protein G | |
RUO | |
1088 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry | |
GM2H6 | |
Unconjugated | |
P31997 | |
CEACAM8 | |
cDNA encoding human CEACAM8 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |