missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A Monoclonal Antibody (SP12), Invitrogen™
Rabbit Monoclonal Antibody
Marca: Invitrogen MA514536
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MA5-14536 targets Chromogranin A in WB, and IHC (P) applications and shows reactivity with Human and Rat samples. This antibody does not cross-react with mouse tissues in IHC applications. The MA5-14536 immunogen is recombinant protein encoding human chromogranin A.
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifica
Chromogranin A | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant diluted in TBS with 1% BSA and 0.1% sodium azide; pH 7.5 | |
P10354, P10645 | |
Chga | |
Recombinant protein encoding human chromogranin A | |
RUO | |
1113, 24258 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
SP12 | |
Unconjugated | |
Chga | |
AL-11; AL26; Beta-granin; betagranin (N-terminal fragment of chromogranin A); catestatin; CgA; CHGA; ChrA; chromofungin; chromogranin A; chromogranin A (parathyroid secretory protein 1); chromogranin A precursor; chromogranin-A; CTNNB; DKFZp686D02253; EA-92; ER-37; ES-43; FLJ25606; FLJ37923; GE-25; GR-44; GV-19; LF-19; LOW QUALITY PROTEIN: chromogranin-A; MGC105435; Pancreastatin; Parastatin; parathyroid secretory protein 1; p-Glu serpinin precursor; pituitary secretory protein I; Serpinin; Serpinin-RRG; SL21; SP-I; SS-18; Vasostatin I; Vasostatin II; Vasostatin-1; Vasostatin-2; WA-8; WE-14 | |
Rabbit | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |