Learn More
DYX1C1, Human, IP-WB Antibody Pair (PW2), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca: Abnova H00161582-PW2.1Set
Includes: Antibody pair for IP: mouse monoclonal anti-DYX1C1 (300μg), Antibody pair for WB: rabbit polyclonal anti-DYX1C1 (50μL)
Specifica
DYX1C1 | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
DYX1/DYXC1/EKN1/FLJ37882/MGC70618/RD | |
Mouse, Rabbit | |
RUO | |
161582 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
DYX1C1 | |
DYX1C1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-DYX1C1 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-DYX1C1 (50 ul) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Titolo del prodotto
For Research Use Only