Learn More
anti-ELF1, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca:  Abnova H00001997-PW1.1Set
Codice prodotto. 16004858
  • 809.00€ / 1 set
Esplora altre offerte speciali
Includes: Antibody pair for IP: rabbit polyclonal anti-ELF1 (300μL), Antibody pair for WB: mouse purified polyclonal anti-ELF1 (50μg)
Questo articolo non è restituibile. Consulta la politica di reso
Specifica
ELF1
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
ELF1
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-ELF1 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-ELF1 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Immunoprecipitation, Western Blot
Unconjugated
ELF1
Mouse, Rabbit
RUO
1997
Human
Liquid
For Research Use Only