Learn More
anti-FGFR2, FGF1 Protein Protein Interaction Antibody Pair, Abnova™
Marca: Abnova DI0198.1Set
Dettagli aggiuntivi : Peso : 0.00010kg
Includes: FGFR2 rabbit purified polyclonal antibody (20μg); FGF1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Descrizione
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the FGFR2 protein, and the other against the FGF1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifica
FGFR2 | |
Monoclonal/Polyclonal | |
FGFR2 | |
FGFR2 | |
1 Set | |
Primary | |
Antibody pair set content: 1. FGFR2 rabbit purified polyclonal antibody (20 ug) 2. FGF1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
BEK/BFR-1/CD332/CEK3/CFD1/ECT1/FLJ98662/JWS/K-SAM/KGFR/TK14/TK25 | |
Mouse, Rabbit | |
RUO | |
2263/2246 | |
Human | |
FGFR2 & FGF1 Protein Protein Interaction Antibody Pair |