missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-HCLS1, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca: Abnova H00003059-PW1.1Set
Includes: Antibody pair for IP: rabbit polyclonal anti-HCLS1 (300μL), Antibody pair for WB: mouse polyclonal anti-HCLS1 (50μL)
Specifica
HCLS1 | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
CTTNL/HS1 | |
Mouse, Rabbit | |
RUO | |
3059 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
HCLS1 | |
HCLS1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-HCLS1 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-HCLS1 (50 ul) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto