missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-HIP1 Ab-1 Matched Antibody Pair, Abnova™
Marca: Abnova H00003092-AP11.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-HIP1 (100μg); Detection antibody: mouse monoclonal anti-HIP1, IgG2a Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
Host Species: Rabbit, Mouse
Isotype: IgG, IgG2a, κ
Cross Reactivity: Human
ELISA Pair
Specifica
HIP1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HIP1. | |
ILWEQ/MGC126506 | |
Mouse, Rabbit | |
1 Set | |
3092 | |
Human | |
IgG, IgG2a |
ELISA | |
Unconjugated | |
HIP1 | |
HIP1 | |
Affinity Purified | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-HIP1 (100 μg) 2. Detection antibody: mouse monoclonal anti-HIP1, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto