missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IL7R Matched Antibody Pair, Abnova™
Marca: Abnova H00003575-AP21.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IL7R (100μg); Detection antibody: mouse purified polyclonal anti-IL7R (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
Host Species: Rabbit, Mouse
Cross Reactivity: Human
ELISA Pair
Specifica
IL7R | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL7R. | |
CD127/CDW127/IL-7R-alpha/IL7RA/ILRA | |
Mouse, Rabbit | |
1 Set | |
3575 | |
Human |
ELISA | |
Unconjugated | |
IL7R | |
IL7R | |
Affinity Purified | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IL7R (100 μg) 2. Detection antibody: mouse purified polyclonal anti-IL7R (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto