missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPD, Human, Matched Antibody Pair (AP43), Abnova™
Marca: Abnova H00006441-AP43.1Set
Includes: Capture antibody: mouse monoclonal anti-SFTPD (100μg); Detection antibody: biotinylated mouse monoclonal anti-SFTPD, IgG1 Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SFTPD.
Host Species: Mouse, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein),ELISA Pair (Transfected Lysate)
Specifica
SFTPD | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SFTPD. | |
COLEC7/PSP-D/SFTP4/SP-D | |
Mouse | |
RUO | |
6441 | |
Human |
ELISA | |
Unconjugated | |
SFTPD | |
SFTPD | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-SFTPD (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-SFTPD, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto