Learn More
SSEA-3 Rat, PE, Clone: MC631, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 560237
Descrizione
The MC631 monoclonal antibody reacts with Stage-Specific Embryonic Antigen-3 (SSEA-3), a carbohydrate epitope on the major ganglioside, but not the neutral glycolipid, of mouse embryos and human teratocarcinoma cells. As its name implies, the expression of SSEA-3 is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. However, its expression pattern differs in the human and mouse. In the human, SSEA-3 is found on teratocarcinoma (embryonal carcinoma or EC), embryonic inner cell mass (ICM), and embryonic stem (ES) cells, and erythrocytes. As human stem cells undergo differentiation, SSEA-3 expression diminishes. In the mouse, SSEA-3 is found on oocytes, ova, zygotes, early cleavage-stage embryos, early blastocysts, ICM, primitive endoderm, and adult kidney, but not on EC or ES cells.
Host Species: Rat
Clone: MC631
Isotype: IgM
Species Reactivity: Human
Immunogen: Mouse Embryos
Flow Cytometry
Specifica
| SSEA-3 | |
| Monoclonal | |
| PE | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| MC631 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse Embryos | |
| 100 Tests | |
| Cell Biology | |
| Human, Mouse | |
| IgM |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.