Learn More
anti-STK11, STRADA Protein Protein Interaction Antibody Pair, Abnova™
Marca: Abnova DI0519.1Set
Dettagli aggiuntivi : Peso : 0.02000kg
Includes: STK11 rabbit purified polyclonal antibody (20μg); STRADA mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Descrizione
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the STK11 protein, and the other against the STRADA protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifica
STK11 | |
Monoclonal/Polyclonal | |
STK11 | |
STK11 | |
1 Set | |
Primary | |
Antibody pair set content: 1. STK11 rabbit purified polyclonal antibody (20 ug) 2. STRADA mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
LKB1/PJS | |
Mouse, Rabbit | |
RUO | |
6794/92335 | |
Human | |
STK11 & STRADA Protein Protein Interaction Antibody Pair |