missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF19, Human, Matched Antibody Pair (AP21), Abnova™
Marca: Abnova H00055504-AP21.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TNFRSF19 (100μg); Detection antibody: mouse polyclonal anti-TNFRSF19 (40μL); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFRSF19.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifica
TNFRSF19 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFRSF19. | |
TAJ/TAJ-alpha/TRADE/TROY | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TNFRSF19 (100 μg) 2. Detection antibody: mouse polyclonal anti-TNFRSF19 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
TNFRSF19 | |
TNFRSF19 | |
Affinity Purified | |
RUO | |
55504 | |
Human |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto